Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps6ka3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212786.1 | NP_997951.1 | 11 - 12 | 1103 | 131 | |
XM_005165118.4 | XP_005165175.1 | 11 - 12 | 1318 | 131 | ||
XM_021476021.1 | XP_021331696.1 | 12 - 13 | 1380 | 131 | ||
GenBank mRNA | BC045856.1 | - | 11 - 12 | 1103 | 131 | |
BC164040.1 | - | 11 - 12 | 941 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |