Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mycla | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212937.1 | NP_998102.1 | 2 - 3 | 678 | 71 | |
GenBank mRNA | BC067723.1 | - | 2 - 3 | 678 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |