Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psma8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213166.1 | NP_998331.1 | 2 - 3 | 327 | 95 | |
GenBank mRNA | BC065608.1 | - | 2 - 3 | 327 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |