Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tha1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002176.1 | NP_001002176.1 | 5 - 6 | 830 | 101 | |
XM_009306814.3 | XP_009305089.1 | 4 - 5 | 769 | 101 | ||
XM_021480050.1 | XP_021335725.1 | 5 - 6 | 575 | 101 | ||
GenBank mRNA | BC072718.1 | - | 5 - 6 | 830 | 101 | |
BC154201.1 | - | 5 - 6 | 816 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |