Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hccsa.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002498.2 | NP_001002498.2 | 5 - 6 | 618 | 97 | |
XM_005163319.4 | XP_005163376.1 | 5 - 6 | 652 | 97 | ||
XM_005163320.4 | XP_005163377.1 | 4 - 5 | 506 | 97 | ||
XM_005163321.4 | XP_005163378.1 | 4 - 5 | 506 | 97 | ||
GenBank mRNA | BC076303.1 | - | 5 - 6 | 618 | 97 | |
BC165701.1 | - | 4 - 5 | 591 | 97 | ||
EH436202.1 | - | 5 - 6 | 644 | 97 | ||
EH462195.1 | - | 2 - 3 | 522 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |