Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aclya | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002649.2 | NP_001002649.2 | 11 - 12 | 1410 | 84 | |
GenBank mRNA | BC076484.1 | - | 11 - 12 | 1348 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |