Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002719.1 | NP_001002719.1 | 5 - 6 | 507 | 136 | |
XM_017356062.2 | XP_017211551.1 | 5 - 6 | 469 | 136 | ||
GenBank mRNA | BC075765.1 | - | 5 - 6 | 507 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |