Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003504.1 | NP_001003504.1 | 7 - 8 | 1046 | 93 | |
GenBank mRNA | BC078339.1 | - | 7 - 8 | 1046 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |