Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013458.2 | NP_001013476.1 | 3 - 4 | 382 | 128 | |
GenBank mRNA | BC091546.1 | - | 3 - 4 | 383 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |