Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018147.1 | NP_001018157.1 | 3 - 4 | 1385 | 119 | |
GenBank mRNA | BC095823.1 | - | 3 - 4 | 1385 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |