Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: olfcw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001018147.1 | NP_001018157.1 | 4 - 5 | 1614 | 76 | |
GenBank mRNA | BC095823.1 | - | 4 - 5 | 1614 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |