Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020627.2 | NP_001018463.1 | 9 - 10 | 1588 | 91 | |
GenBank mRNA | BC095378.1 | - | 9 - 10 | 1590 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |