Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020772.1 | NP_001018608.1 | 4 - 5 | 669 | 105 | |
XM_009292818.3 | XP_009291093.1 | 4 - 5 | 909 | 105 | ||
XM_009292819.3 | XP_009291094.1 | 4 - 5 | 730 | 143 | ||
XM_017351638.2 | XP_017207127.1 | 4 - 5 | 730 | 105 | ||
XM_017351639.2 | XP_017207128.1 | 4 - 5 | 730 | 143 | ||
XM_021467167.1 | XP_021322842.1 | 4 - 5 | 598 | 105 | ||
GenBank mRNA | BC095858.1 | - | 4 - 5 | 669 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |