Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afg1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025482.2 | NP_001020653.2 | 10 - 11 | 1140 | 126 | |
XM_021468423.1 | XP_021324098.1 | 9 - 10 | 840 | 126 | ||
GenBank mRNA | BC115066.1 | - | 10 - 11 | 1140 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |