Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afg1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025482.2 | NP_001020653.2 | 12 - 13 | 1396 | 83 | |
XM_021468423.1 | XP_021324098.1 | 11 - 12 | 83 | |||
GenBank mRNA | BC115066.1 | - | 12 - 13 | 1396 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |