Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afg1lb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025482.2 | NP_001020653.2 | 4 - 5 | 604 | 150 | |
XM_021468423.1 | XP_021324098.1 | 3 - 4 | 304 | 150 | ||
GenBank mRNA | BC115066.1 | - | 4 - 5 | 604 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |