Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apobb.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030062.1 | NP_001025233.1 | 19 - 20 | 3190 | 97 | |
XM_005160629.3 | XP_005160686.1 | 20 - 21 | 3293 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |