Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: paqr3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030148.2 | NP_001025319.2 | 5 - 6 | 881 | 142 | |
XM_005155617.4 | XP_005155674.1 | 5 - 6 | 864 | 142 | ||
GenBank mRNA | BC096792.1 | - | 5 - 6 | 881 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |