Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gigyf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030161.2 | NP_001025332.2 | 21 - 22 | 2649 | 78 | |
XM_017358654.2 | XP_017214143.1 | 21 - 22 | 2740 | 78 | ||
GenBank mRNA | BC098603.1 | - | 21 - 22 | 2667 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |