Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: emilin1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030207.1 | NP_001025378.1 | 6 - 7 | 2700 | 124 | |
XM_005160756.3 | XP_005160813.1 | 7 - 8 | 3058 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Danio rerio Probe spans exons |