Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3hb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035264.2 | NP_001030341.1 | 2 - 3 | 282 | 68 | |
GenBank mRNA | BC086809.1 | - | 2 - 3 | 248 | 68 | |
BC107824.1 | - | 2 - 3 | 283 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |