Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ntn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037415.1 | NP_001032492.1 | 3 - 4 | 1434 | 54 | |
GenBank mRNA | AY928518.1 | - | 3 - 4 | 1434 | 54 | |
KM655684.1 | - | 1 - 2 | 1029 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Danio rerio Probe spans exons |