Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wt1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039634.2 | NP_001034723.2 | 2 - 3 | 673 | 74 | |
XM_005159243.4 | XP_005159300.1 | 2 - 3 | 946 | 74 | ||
GenBank mRNA | BC124087.1 | - | 2 - 3 | 673 | 74 | |
BC164718.1 | - | 2 - 3 | 531 | 74 | ||
DQ145942.1 | - | 2 - 3 | 497 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |