Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abcg2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039639.2 | NP_001034728.2 | 8 - 9 | 967 | 153 | |
XM_005156523.4 | XP_005156580.1 | 8 - 9 | 1079 | 153 | ||
XM_009307089.3 | XP_009305364.1 | 8 - 9 | 1148 | 153 | ||
GenBank mRNA | BC124086.1 | - | 8 - 9 | 967 | 153 | |
BC165778.1 | - | 7 - 8 | 946 | 153 | ||
DQ223860.1 | - | 7 - 8 | 912 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |