Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: specc1la | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039816.1 | NP_001034905.1 | 2 - 3 | 317 | 65 | |
XM_005167177.4 | XP_005167234.1 | 3 - 4 | 509 | 65 | ||
XM_009304055.3 | XP_009302330.1 | 3 - 4 | 509 | 65 | ||
GenBank mRNA | AY884302.1 | - | 2 - 3 | 317 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Danio rerio Probe spans exons |