Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hapln4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044907.1 | NP_001038372.1 | 3 - 4 | 886 | 105 | |
XM_005170459.4 | XP_005170516.1 | 4 - 5 | 1096 | 105 | ||
GenBank mRNA | BC142773.1 | - | 2 - 3 | 958 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |