Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tiparp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045019.3 | NP_001038484.1 | 3 - 4 | 1196 | 85 | |
GenBank mRNA | EE311950.1 | - | 2 - 3 | 416 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |