Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tiparp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045019.3 | NP_001038484.1 | 4 - 5 | 1380 | 73 | |
GenBank mRNA | CA472479.1 | - | 3 - 4 | 499 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |