Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhrs12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076557.1 | NP_001070025.1 | 7 - 8 | 619 | 63 | |
GenBank mRNA | BC122377.1 | - | 7 - 8 | 619 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |