Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: isoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079981.1 | NP_001073450.1 | 4 - 5 | 726 | 66 | |
GenBank mRNA | BC124430.1 | - | 4 - 5 | 726 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |