Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taar12h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082909.1 | NP_001076378.1 | 1 | 747 | 76 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |