Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sin3aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098180.1 | NP_001091650.1 | 9 - 10 | 1570 | 58 | |
XM_005162970.4 | XP_005163027.1 | 9 - 10 | 1529 | 58 | ||
GenBank mRNA | BC139856.1 | - | 9 - 10 | 1570 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |