Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099994.2 | NP_001093464.1 | 4 - 5 | 473 | 86 | |
XM_009298589.3 | XP_009296864.1 | 4 - 5 | 523 | 86 | ||
XM_021480548.1 | XP_021336223.1 | 4 - 5 | 531 | 86 | ||
GenBank mRNA | BC117659.1 | - | 4 - 5 | 468 | 86 | |
BC171413.1 | - | 4 - 5 | 465 | 86 | ||
BC171415.1 | - | 4 - 5 | 465 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |