Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mntb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103111.1 | NP_001096581.1 | 3 - 4 | 896 | 129 | |
GenBank mRNA | BC150227.1 | - | 3 - 4 | 896 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |