Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mitfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130923.2 | NP_570998.1 | 5 - 6 | 647 | 99 | |
XM_021476775.1 | XP_021332450.1 | 6 - 7 | 822 | 99 | ||
GenBank mRNA | AF119220.1 | - | 5 - 6 | 647 | 99 | |
BC056318.1 | - | 5 - 6 | 639 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |