Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mitfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001362262.1 | NP_001349191.1 | 9 - 10 | 984 | 90 | |
NM_130923.2 | NP_570998.1 | 6 - 7 | 696 | 90 | ||
XM_021476774.1 | XP_021332449.1 | 6 - 7 | 758 | 90 | ||
XM_021476775.1 | XP_021332450.1 | 7 - 8 | 871 | 90 | ||
GenBank mRNA | AF119220.1 | - | 6 - 7 | 696 | 90 | |
BC056318.1 | - | 6 - 7 | 688 | 90 | ||
BC065647.1 | - | 6 - 7 | 716 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |