Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mitfa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001362262.1 | NP_001349191.1 | 11 - 12 | 1205 | 65 | |
NM_130923.2 | NP_570998.1 | 8 - 9 | 917 | 65 | ||
XM_021476774.1 | XP_021332449.1 | 8 - 9 | 979 | 65 | ||
XM_021476775.1 | XP_021332450.1 | 9 - 10 | 1092 | 65 | ||
GenBank mRNA | AF119220.1 | - | 8 - 9 | 917 | 65 | |
BC056318.1 | - | 8 - 9 | 909 | 65 | ||
BC065647.1 | - | 8 - 9 | 937 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |