Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lft2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130961.1 | NP_571036.1 | 2 - 3 | 519 | 65 | |
GenBank mRNA | AF132445.1 | - | 2 - 3 | 519 | 65 | |
BC097092.1 | - | 2 - 3 | 543 | 65 | ||
KM655581.1 | - | 2 - 3 | 510 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |