Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130973.3 | NP_571048.2 | 6 - 7 | 852 | 67 | |
GenBank mRNA | AF034606.1 | - | 6 - 7 | 67 | ||
BC162594.1 | - | 6 - 7 | 854 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |