Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ddx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131057.1 | NP_571132.1 | 23 - 24 | 1647 | 77 | |
XM_005156453.4 | XP_005156510.1 | 23 - 24 | 1659 | 77 | ||
XM_017357926.2 | XP_017213415.1 | 23 - 24 | 1656 | 77 | ||
XM_017357927.2 | XP_017213416.1 | 22 - 23 | 1626 | 77 | ||
XM_021479102.1 | XP_021334777.1 | 22 - 23 | 1623 | 77 | ||
XM_021479103.1 | XP_021334778.1 | 21 - 22 | 1526 | 77 | ||
XM_021479104.1 | XP_021334779.1 | 21 - 22 | 1570 | 77 | ||
XM_021479105.1 | XP_021334780.1 | 20 - 21 | 1490 | 77 | ||
GenBank mRNA | AB005147.1 | - | 23 - 24 | 1647 | 77 | |
BC129275.1 | - | 21 - 22 | 1578 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |