Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: her1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131078.1 | NP_571153.1 | 2 - 3 | 195 | 147 | |
GenBank mRNA | BC162281.1 | - | 2 - 3 | 160 | 147 | |
X97329.1 | - | 195 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |