Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131226.2 | NP_571301.2 | 3 - 4 | 892 | 56 | |
GenBank mRNA | AF001908.1 | - | 2 - 3 | 541 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |