Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ephb4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131417.1 | NP_571492.1 | 2 - 3 | 153 | 91 | |
GenBank mRNA | AJ005029.1 | - | 2 - 3 | 153 | 91 | |
BC163275.1 | - | 2 - 3 | 91 | |||
KM655609.1 | - | 2 - 3 | 153 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |