Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131480.1 | NP_571555.1 | 16 - 17 | 1527 | 114 | |
GenBank mRNA | AJ005692.1 | - | 16 - 17 | 1527 | 114 | |
BC045863.1 | - | 15 - 16 | 1674 | 114 | ||
BC060933.1 | - | 16 - 17 | 1556 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |