Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csf1ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131672.1 | NP_571747.1 | 6 - 7 | 1228 | 82 | |
XM_021481056.1 | XP_021336731.1 | 7 - 8 | 1358 | 82 | ||
XM_021481057.1 | XP_021336732.1 | 7 - 8 | 1285 | 82 | ||
GenBank mRNA | AF240639.1 | - | 6 - 7 | 1228 | 82 | |
BC162803.1 | - | 6 - 7 | 1163 | 82 | ||
CU458899.1 | - | 6 - 7 | 1122 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |