Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131801.2 | NP_571876.1 | 9 - 10 | 1322 | 87 | |
GenBank mRNA | AB069857.1 | - | 9 - 10 | 1322 | 87 | |
AF290981.1 | - | 8 - 9 | 1187 | 87 | ||
BC047796.1 | - | 8 - 9 | 1287 | 87 | ||
BC063932.1 | - | 9 - 10 | 1293 | 87 | ||
BC097034.1 | - | 8 - 9 | 1400 | 87 | ||
BC165342.1 | - | 7 - 8 | 1059 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |