Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: meis1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131893.2 | NP_571968.1 | 3 - 4 | 1011 | 124 | |
GenBank mRNA | AF375871.2 | - | 3 - 4 | 758 | 124 | |
BC060891.1 | - | 3 - 4 | 534 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |