Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sf3b4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153661.4 | NP_705947.3 | 3 - 4 | 738 | 82 | |
GenBank mRNA | AY099515.2 | - | 3 - 4 | 729 | 82 | |
BC056532.1 | - | 3 - 4 | 724 | 82 | ||
BC067655.1 | - | 3 - 4 | 728 | 82 | ||
CT611775.2 | - | 2 - 3 | 658 | 82 | ||
EH501773.1 | - | 3 - 4 | 760 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |