Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lamb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173275.1 | NP_775382.1 | 15 - 16 | 1846 | 103 | |
GenBank mRNA | AF468049.1 | - | 15 - 16 | 1846 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |