Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arntl1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178300.1 | NP_840085.1 | 12 - 13 | 1329 | 82 | |
XM_005166616.4 | XP_005166673.1 | 15 - 16 | 1587 | 82 | ||
XM_009303573.3 | XP_009301848.1 | 14 - 15 | 1518 | 82 | ||
XM_021477412.1 | XP_021333087.1 | 14 - 15 | 1525 | 82 | ||
GenBank mRNA | AB087257.1 | - | 12 - 13 | 1329 | 82 | |
BC134895.1 | - | 15 - 16 | 1572 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |